Gene Transcripts

Gene Symbol
Tmem199
Entrez Gene ID
195040
Gene Name
transmembrane protein 199
Gene Aliases
AI316787, AI848469, R74819
Chromosome Location
Chr. 11: 78507055 - 78512168 on Build GRCm38
UniGene ID
Mm.383431
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199199.3 NP_954669.2 6 921
GenBank mRNA AK160676.1 - 1 327
BC050977.1 - 6 884
BC064015.1 - 2 697

View Full Product Details