Gene Transcripts

Gene Symbol
Tmem200a
Entrez Gene ID
77220
Gene Name
transmembrane protein 200A
Gene Aliases
C030003D03Rik
Chromosome Location
Chr. 10: 25991182 - 26079408 on Build GRCm38
UniGene ID
Mm.266260
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029881.3 NP_084157.2 2 3549
GenBank mRNA AK032569.1 - 2 3148
AK048673.1 - 3 2944
AK165025.1 - 2 2717
AK173299.1 - 2 3549

View Full Product Details