Gene Transcripts
- Gene Symbol
- Gm6793
- Entrez Gene ID
- 627828
- Gene Name
- heterogeneous nuclear ribonucleoprotein A3 pseudogene
- Gene Aliases
- EG627828
- Chromosome Location
- Chr. 8: 112013433 - 112015036 on Build GRCm38
- UniGene ID
- Mm.459502
- Species
- Mus musculus
- Gene Symbol
- Hnrnpa3
- Entrez Gene ID
- 229279
- Gene Name
- heterogeneous nuclear ribonucleoprotein A3
- Gene Aliases
- 2410013L13Rik, 2610209F03Rik, 2610510D13Rik, Hnrpa3
- Chromosome Location
- Chr. 2: 75659230 - 75669407 on Build GRCm38
- UniGene ID
- Mm.316306
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053263.1 |
NP_444493.1
|
8, 9 |
954 |
|
NM_146130.3 |
NP_666242.2
|
8, 9 |
1020 |
|
NM_198090.2 |
NP_932758.1
|
8, 9 |
1020 |
|
GenBank mRNA |
AB201711.1 |
- |
8, 9 |
1000 |
|
AF463524.1 |
- |
8, 9 |
932 |
|
AK133860.1 |
- |
8, 9 |
1019 |
|
AK169663.1 |
- |
8, 9 |
1002 |
|
BC023828.1 |
- |
8, 9 |
973 |
5343619 |
BC023908.1 |
- |
8, 9 |
989 |
5322033 |
BC024454.1 |
- |
5, 6 |
537 |
|
BC038364.1 |
- |
8, 9 |
1002 |
5362605 |
BC062198.1 |
- |
8, 9 |
828 |
2655879 |
BC064824.1 |
- |
8, 9 |
985 |
6477672 |
BC085474.1 |
- |
7, 8 |
895 |
30538435 |
BC092083.1 |
- |
5, 6 |
615 |
|
BC116250.1 |
- |
8, 9 |
923 |
40093600 |
BC116251.1 |
- |
8, 9 |
923 |
40093602 |
BC157919.1 |
- |
8, 9 |
923 |
9007347 |
BC158038.1 |
- |
8, 9 |
923 |
9007341 |
View Full Product Details