Gene Transcripts
- Gene Symbol
- GABRA5
- Entrez Gene ID
- 2558
- Gene Name
- gamma-aminobutyric acid type A receptor alpha5 subunit
- Gene Aliases
- Chromosome Location
- Chr. 15: 26866719 - 26949210 on Build GRCh38
- UniGene ID
- Hs.24969
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000810.3 |
NP_000801.1
|
8, 9 |
1264 |
|
NM_001165037.1 |
NP_001158509.1
|
8, 9 |
1130 |
|
XM_005268258.2 |
XP_005268315.1
|
8, 9 |
1142 |
|
XM_006720459.2 |
XP_006720522.1
|
8, 9 |
1046 |
|
XM_017022055.1 |
XP_016877544.1
|
8, 9 |
1012 |
|
XM_017022056.1 |
XP_016877545.1
|
8, 9 |
1046 |
|
GenBank mRNA |
A28104.1 |
- |
8, 9 |
1029 |
|
AK290453.1 |
- |
8, 9 |
927 |
|
AK294303.1 |
- |
5, 6 |
587 |
|
AK303742.1 |
- |
3, 4 |
435 |
|
BC111979.1 |
- |
7, 8 |
876 |
8327447 |
BC113422.1 |
- |
7, 8 |
876 |
8322474 |
BT009830.1 |
- |
6, 7 |
732 |
|
L08485.1 |
- |
8, 9 |
1037 |
|
View Full Product Details