Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018130.2 | NP_060600.2 | 4 | 466 | |
XM_011533895.2 | XP_011532197.1 | 4 | 494 | ||
XM_011533896.2 | XP_011532198.1 | 4 | 494 | ||
XM_011533897.2 | XP_011532199.1 | 4 | 494 | ||
XM_011533898.2 | XP_011532200.1 | 4 | 494 | ||
XM_011533899.1 | XP_011532201.1 | 4 | 494 | ||
XM_017006724.1 | XP_016862213.1 | 4 | 494 | ||
GenBank mRNA | AK001401.1 | - | 4 | 464 | |
AK092962.1 | - | 4 | 464 | ||
AK296789.1 | - | 4 | 365 | ||
BC017204.2 | - | 4 | 456 | 3617948 | |
BC017274.2 | - | 4 | 444 | 4840343 | |
BC025270.1 | - | 4 | 456 | 4622871 | |
BC032671.1 | - | 4 | 470 | 5538687 | |
BC039830.1 | - | 4 | 399 | 6064610 | |
BC047879.2 | - | 4 | 417 | 5541019 |