Gene Transcripts

Gene Symbol
FAM170B
Entrez Gene ID
170370
Gene Name
family with sequence similarity 170 member B
Gene Aliases
C10orf73
Chromosome Location
Chr. 10: 49131154 - 49134020 on Build GRCh38
UniGene ID
Hs.646291
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164484.1 NP_001157956.1 2 1234
GenBank mRNA BC029839.1 - 2 1222
BC047597.1 - 2 1234

View Full Product Details