Gene Transcripts
- Gene Symbol
- LOC283278
- Entrez Gene ID
- 283278
- Gene Name
- pleckstrin homology domain-containing family A member 6
- Gene Aliases
- Chromosome Location
- Chr. 11: 16778935 - 16787192 on Build GRCh38
- UniGene ID
- Hs.729280
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL833334.1 |
- |
1 |
1867 |
|
AL833346.1 |
- |
1 |
1867 |
|
View Full Product Details