Gene Transcripts

Gene Symbol
ARL16
Entrez Gene ID
339231
Gene Name
ADP ribosylation factor like GTPase 16
Gene Aliases
Chromosome Location
Chr. 17: 81681194 - 81683924 on Build GRCh38
UniGene ID
Hs.658789
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040025.1 NP_001035114.1 5 857
GenBank mRNA AK095847.1 - 3 1985
AK304718.1 - 5 696
BC042380.1 - 5 695
BC063687.1 - 5 659

View Full Product Details