Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001256703.1 | NP_001243632.1 | 3 | 724 | |
NM_001256704.1 | NP_001243633.1 | 5 | 877 | ||
NM_001267582.1 | NP_001254511.1 | 6 | 545 | ||
NM_002042.4 | NP_002033.2 | 4 | 775 | ||
XM_017010689.1 | XP_016866178.1 | 5 | 365 | ||
GenBank mRNA | AK296124.1 | - | 3 | 316 | |
AK298948.1 | - | 6 | 535 | ||
AK313609.1 | - | 5 | 877 | ||
BC130344.1 | - | 4 | 329 | 40146375 | |
DA753437.1 | - | 5 | 471 | ||
M62400.1 | - | 4 | 348 |