Gene Transcripts
- Gene Symbol
- LOC105376351
- Entrez Gene ID
- 105376351
- Gene Name
- uncharacterized LOC105376351
- Gene Aliases
- Chromosome Location
- Chr. 10: 2446253 - 2501462 on Build GRCh38
- UniGene ID
- Hs.343482
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_136146.1 |
|
3 |
478 |
|
View Full Product Details