Gene Transcripts
- Gene Symbol
- AGBL1-AS1
- Entrez Gene ID
- 727915
- Gene Name
- AGBL1 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 15: 86296833 - 86316968 on Build GRCh38
- UniGene ID
- Hs.451281
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046012.1 |
|
1 |
100 |
|
GenBank mRNA |
CD359326.1 |
- |
1 |
99 |
|
DB459014.1 |
- |
1 |
305 |
|
View Full Product Details