Gene Transcripts

Gene Symbol
PRAMEF12
Entrez Gene ID
390999
Gene Name
PRAME family member 12
Gene Aliases
Chromosome Location
Chr. 1: 12774848 - 12777906 on Build GRCh38
UniGene ID
Hs.156406
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080830.4 NP_001074299.2 2 777

View Full Product Details