Gene Transcripts
- Gene Symbol
- RBM41
- Entrez Gene ID
- 55285
- Gene Name
- RNA binding motif protein 41
- Gene Aliases
- Chromosome Location
- Chr. X: 107061885 - 107118827 on Build GRCh38
- UniGene ID
- Hs.139053
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001171080.1 |
NP_001164551.1
|
5, 6 |
989 |
|
NM_001324242.1 |
NP_001311171.1
|
6, 7 |
1061 |
|
NM_001324243.1 |
NP_001311172.1
|
6, 7 |
1154 |
|
NM_001324244.1 |
NP_001311173.1
|
6, 7 |
1061 |
|
NM_018301.4 |
NP_060771.3
|
5, 6 |
989 |
|
NR_136728.1 |
|
5, 6 |
657 |
|
NR_136729.1 |
|
6, 7 |
751 |
|
NR_136732.1 |
|
5, 6 |
750 |
|
XM_011530980.2 |
XP_011529282.1
|
6, 7 |
1153 |
|
XM_011530981.2 |
XP_011529283.1
|
5, 6 |
1081 |
|
GenBank mRNA |
AK001878.1 |
- |
5, 6 |
962 |
|
BC006986.1 |
- |
5, 6 |
966 |
3995903 |
View Full Product Details