Gene Transcripts
- Gene Symbol
- LOC105371226
- Entrez Gene ID
- 105371226
- Gene Name
- uncharacterized LOC105371226
- Gene Aliases
- Chromosome Location
- Chr. 1: 148271883 - 148282357 on Build GRCh38
- UniGene ID
- Hs.263424
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK096567.1 |
- |
1 |
169 |
|
View Full Product Details