Gene Transcripts

Gene Symbol
LINC00608
Entrez Gene ID
151300
Gene Name
long intergenic non-protein coding RNA 608
Gene Aliases
Chromosome Location
Chr. 2: 218976284 - 218977922 on Build GRCh38
UniGene ID
Hs.420716
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015390.1 2 786
NR_024385.1 2 1262
GenBank mRNA AK098539.1 - 2 1262
BC033552.1 - 2 1244
BC049373.1 - 2 786

View Full Product Details