Gene Transcripts

Gene Symbol
LINC00494
Entrez Gene ID
284749
Gene Name
long intergenic non-protein coding RNA 494
Gene Aliases
Chromosome Location
Chr. 20: 48359911 - 48370638 on Build GRCh38
UniGene ID
Hs.299080
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026958.1 5 2016
GenBank mRNA AL832259.1 - 3 1504
BC035164.2 - 5 1980
BC122555.1 - 1 385 40084813

View Full Product Details