Gene Transcripts

Gene Symbol
TMEM247
Entrez Gene ID
388946
Gene Name
transmembrane protein 247
Gene Aliases
Chromosome Location
Chr. 2: 46479565 - 46484425 on Build GRCh38
UniGene ID
Hs.733348
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145051.2 NP_001138523.1 3 598

View Full Product Details