Gene Transcripts

Gene Symbol
FAM209A
Entrez Gene ID
200232
Gene Name
family with sequence similarity 209 member A
Gene Aliases
C20orf106, dJ1153D9.3
Chromosome Location
Chr. 20: 56524729 - 56533590 on Build GRCh38
UniGene ID
Hs.504907
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012971.3 NP_001012989.2 2 448
GenBank mRNA BC070179.1 - 2 410
BC107704.1 - 2 423 6614835
BX335293.2 - 2 448

View Full Product Details