Gene Transcripts
- Gene Symbol
- PP7080
- Entrez Gene ID
- 25845
- Gene Name
- uncharacterized LOC25845
- Gene Aliases
- Chromosome Location
- Chr. 5: 470510 - 472965 on Build GRCh38
- UniGene ID
- Hs.719132
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024158.1 |
|
2 |
1713 |
|
GenBank mRNA |
AF370427.1 |
- |
2 |
1814 |
|
BC086861.1 |
- |
2 |
1694 |
|
View Full Product Details