Gene Transcripts

Gene Symbol
PP7080
Entrez Gene ID
25845
Gene Name
uncharacterized LOC25845
Gene Aliases
Chromosome Location
Chr. 5: 470510 - 472965 on Build GRCh38
UniGene ID
Hs.719132
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024158.1 2 1176
GenBank mRNA AF370427.1 - 2 1276
BC045808.1 - 2 1015
BC072677.1 - 2 1030
BC086861.1 - 2 1157

View Full Product Details