Gene Transcripts

Gene Symbol
Tefm
Entrez Gene ID
287554
Gene Name
transcription elongation factor, mitochondrial
Gene Aliases
RGD1306595
Chromosome Location
Chr. 10: 67476006 - 67478830 on Build Rnor_6.0
UniGene ID
Rn.16586
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025626.1 NP_001020797.1 3 1076
GenBank mRNA BC098792.1 - 3 1076

View Full Product Details