Gene Transcripts

Gene Symbol
Cpsf3
Entrez Gene ID
298916
Gene Name
cleavage and polyadenylation specific factor 3, 73kDa
Gene Aliases
Chromosome Location
Chr. 6: 43363792 - 43391712 on Build Rnor_6.0
UniGene ID
Rn.100522
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001030030.1 NP_001025201.1 10, 11 1332
GenBank mRNA BC099817.1 - 10, 11 1332

View Full Product Details