Gene Transcripts

Gene Symbol
Rexo4
Entrez Gene ID
311826
Gene Name
REX4, RNA exonuclease 4 homolog (S. cerevisiae)
Gene Aliases
Xpmc2h
Chromosome Location
Chr. 3: 5500578 - 5510908 on Build Rnor_6.0
UniGene ID
Rn.116879
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033884.1 NP_001029056.1 8 2293
GenBank mRNA BC161820.1 - 8 2258

View Full Product Details