Gene Transcripts

Gene Symbol
Akirin1
Entrez Gene ID
595134
Gene Name
akirin 1
Gene Aliases
Chromosome Location
Chr. 5: 141415226 - 141430627 on Build Rnor_6.0
UniGene ID
Rn.3674
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001030054.1 NP_001025225.1 5 1437
GenBank mRNA CO384190.1 - 1 347

View Full Product Details