Gene Transcripts

Gene Symbol
Olfr1436
Entrez Gene ID
258682
Gene Name
olfactory receptor 1436
Gene Aliases
MOR214-2
Chromosome Location
Chr. 19: 12298183 - 12299130 on Build GRCm38
UniGene ID
Mm.377636
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146687.2 NP_666898.2 1 778
GenBank mRNA BC127991.1 - 1 782

View Full Product Details