Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001318781.1 | NP_001305710.1 | 14 | 2225 | |
NM_182920.1 | NP_891550.1 | 15 | 2309 | ||
GenBank mRNA | AB037733.1 | - | 14 | 1804 | |
AF261918.1 | - | 15 | 2289 | ||
AF488803.1 | - | 15 | 2309 | ||
BC130578.1 | - | 15 | 2299 | 40146609 | |
BC171764.1 | - | 14 | 2215 | 9054417 |