Gene Transcripts

Gene Symbol
CASS4
Entrez Gene ID
57091
Gene Name
Cas scaffolding protein family member 4
Gene Aliases
C20orf32, CAS4, HEFL, HEPL
Chromosome Location
Chr. 20: 56411550 - 56460240 on Build GRCh38
UniGene ID
Hs.473144
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164114.1 NP_001157586.1 5 1649
NM_001164116.1 NP_001157588.1 6 1835
NM_020356.3 NP_065089.2 5 1811
XM_006723831.3 XP_006723894.1 6 2235
GenBank mRNA AJ276678.1 - 6 1832
AK295613.1 - 5 1514
AK308057.1 - 5 1812
AK314653.1 - 5 1744
BC027951.1 - 5 1761 5205865

View Full Product Details