Gene Transcripts

Gene Symbol
Miip
Entrez Gene ID
298643
Gene Name
migration and invasion inhibitory protein
Gene Aliases
Iip45
Chromosome Location
Chr. 5: 164672464 - 164679503 on Build Rnor_6.0
UniGene ID
Rn.27362
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017450.1 NP_001017450.1 10 1524
XM_006239379.2 XP_006239441.1 10 1599
XM_006239380.2 XP_006239442.1 10 1697
XM_006239381.2 XP_006239443.1 10 1626
GenBank mRNA BC079047.1 - 10 1524
FJ618907.1 - 10 1728
FQ227320.1 - 10 1482

Gene Symbol
LOC100911456
Entrez Gene ID
100911456
Gene Name
migration and invasion-inhibitory protein-like
Gene Aliases
Chromosome Location
Chr. 5: 164548657 - 164555842 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006239406.2 XP_006239468.1 1599
XM_006239407.2 XP_006239469.1 1697
XM_006239408.2 XP_006239470.1 1723
XM_006239409.2 XP_006239471.1 1626

View Full Product Details