Gene Transcripts

Gene Symbol
Olfr1290
Entrez Gene ID
257662
Gene Name
olfactory receptor 1290
Gene Aliases
MOR248-9
Chromosome Location
Chr. 2: 111489218 - 111493815 on Build GRCm38
UniGene ID
Mm.484725
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278787.1 NP_001265716.1 2 882
NM_001278788.1 NP_001265717.1 2 875
NM_146262.3 NP_666374.2 3 1091

View Full Product Details