Gene Transcripts

Gene Symbol
DDRGK1
Entrez Gene ID
65992
Gene Name
DDRGK domain containing 1
Gene Aliases
C20orf116, UFBP1, dJ1187M17.3
Chromosome Location
Chr. 20: 3190350 - 3204697 on Build GRCh38
UniGene ID
Hs.471975
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023935.2 NP_076424.1 9 1210
GenBank mRNA BC000643.1 - 9 1162

View Full Product Details