Gene Transcripts
- Gene Symbol
- RHBDF2
- Entrez Gene ID
- 79651
- Gene Name
- rhomboid 5 homolog 2
- Gene Aliases
- RHBDL5, RHBDL6, TEC, TOC, TOCG, iRhom2
- Chromosome Location
- Chr. 17: 76470893 - 76501440 on Build GRCh38
- UniGene ID
- Hs.464157
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001005498.3 |
NP_001005498.2
|
6, 7 |
1028 |
|
NM_024599.5 |
NP_078875.4
|
7, 8 |
1190 |
|
XM_005257669.3 |
XP_005257726.1
|
7, 8 |
1551 |
|
XM_005257670.1 |
XP_005257727.1
|
7, 8 |
1060 |
|
XM_011525249.2 |
XP_011523551.1
|
7, 8 |
2507 |
|
XM_011525250.2 |
XP_011523552.1
|
8, 9 |
1217 |
|
XM_011525251.2 |
XP_011523553.1
|
7, 8 |
1034 |
|
XM_017025079.1 |
XP_016880568.1
|
7, 8 |
1464 |
|
GenBank mRNA |
AK025994.1 |
- |
2, 3 |
214 |
|
AK057882.1 |
- |
4, 5 |
884 |
|
AK074034.1 |
- |
5, 6 |
1495 |
|
AK225684.1 |
- |
2, 3 |
167 |
|
AK292135.1 |
- |
7, 8 |
1183 |
|
AK292166.1 |
- |
6, 7 |
983 |
|
AK303434.1 |
- |
5, 6 |
844 |
|
AL833680.1 |
- |
1, 2 |
83 |
|
AY500247.1 |
- |
6, 7 |
879 |
|
BC016034.1 |
- |
7, 8 |
1170 |
3910543 |
BC035829.1 |
- |
2, 3 |
276 |
|
View Full Product Details