Gene Transcripts
- Gene Symbol
- ACAP3
- Entrez Gene ID
- 116983
- Gene Name
- ArfGAP with coiled-coil, ankyrin repeat and PH domains 3
- Gene Aliases
- CENTB5
- Chromosome Location
- Chr. 1: 1292384 - 1307925 on Build GRCh38
- UniGene ID
- Hs.535257
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_030649.2 |
NP_085152.2
|
5, 6 |
421 |
|
XM_005244715.2 |
XP_005244772.1
|
5, 6 |
457 |
|
XM_011540606.2 |
XP_011538908.1
|
5, 6 |
470 |
|
XM_011540607.1 |
XP_011538909.1
|
5, 6 |
2894 |
|
XM_011540608.2 |
XP_011538910.1
|
5, 6 |
471 |
|
XM_011540609.2 |
XP_011538911.1
|
5, 6 |
471 |
|
GenBank mRNA |
AB051503.1 |
- |
4, 5 |
2086 |
|
AF411981.1 |
- |
3, 4 |
221 |
|
AK093954.1 |
- |
5, 6 |
468 |
|
BC051194.1 |
- |
5, 6 |
421 |
4819254 |
View Full Product Details