Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_032290.3 | NP_115666.2 | 16 | 2137 | |
XM_017009978.1 | XP_016865467.1 | 15, 16 | 2246 | ||
XM_017009979.1 | XP_016865468.1 | 16 | 6899 | ||
XM_017009980.1 | XP_016865469.1 | 15 | 2169 | ||
XM_017009981.1 | XP_016865470.1 | 15, 16 | 2097 | ||
XM_017009982.1 | XP_016865471.1 | 16 | 2318 | ||
XM_017009983.1 | XP_016865472.1 | 6 | 780 | ||
XM_017009984.1 | XP_016865473.1 | 9, 10 | 1171 | ||
GenBank mRNA | AK299080.1 | - | 16 | 2163 | |
AK309859.1 | - | 3 | 333 | ||
AL136560.1 | - | 2 | 169 | ||
BC063674.1 | - | 2 | 639 | 4368392 | |
CR533539.1 | - | 2 | 149 | ||
CR936768.1 | - | 16 | 2143 |