Gene Transcripts

Gene Symbol
ZNF80
Entrez Gene ID
7634
Gene Name
zinc finger protein 80
Gene Aliases
pT17
Chromosome Location
Chr. 3: 114211035 - 114237578 on Build GRCh38
UniGene ID
Hs.271079
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007136.3 NP_009067.2 1 588
XM_017007132.1 XP_016862621.1 1 276
XM_017007133.1 XP_016862622.1 1 276
XM_017007134.1 XP_016862623.1 1 276
XM_017007135.1 XP_016862624.1 1 276
XM_017007136.1 XP_016862625.1 1 276
GenBank mRNA BC069606.1 - 1 282
BC069826.1 - 1 281 7262419
BC129925.1 - 1 111 40121991
X65233.1 - 1 596

View Full Product Details