Gene Transcripts

Gene Symbol
C19orf18
Entrez Gene ID
147685
Gene Name
chromosome 19 open reading frame 18
Gene Aliases
Chromosome Location
Chr. 19: 57958437 - 57974534 on Build GRCh38
UniGene ID
Hs.134209
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152474.4 NP_689687.1 1, 2 222
GenBank mRNA BC033933.2 - 1, 2 225

View Full Product Details