Gene Transcripts

Gene Symbol
FBXL14
Entrez Gene ID
144699
Gene Name
F-box and leucine rich repeat protein 14
Gene Aliases
Fbl14
Chromosome Location
Chr. 12: 1565993 - 1594189 on Build GRCh38
UniGene ID
Hs.367956
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152441.2 NP_689654.1 2 1912
XM_017018875.1 XP_016874364.1 2 1894
GenBank mRNA BC028132.1 - 2 1910

View Full Product Details