Gene Transcripts
- Gene Symbol
- TMEM86A
- Entrez Gene ID
- 144110
- Gene Name
- transmembrane protein 86A
- Gene Aliases
- Chromosome Location
- Chr. 11: 18698735 - 18704785 on Build GRCh38
- UniGene ID
- Hs.502100
- Species
- Homo sapiens
- Gene Symbol
- IGSF22
- Entrez Gene ID
- 283284
- Gene Name
- immunoglobulin superfamily member 22
- Gene Aliases
- IGFN2
- Chromosome Location
- Chr. 11: 18704305 - 18726939 on Build GRCh38
- UniGene ID
- Hs.434152
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AA255875.1 |
- |
1 |
44 |
|
AI952274.1 |
- |
|
47 |
|
View Full Product Details