Gene Transcripts

Gene Symbol
HOXD10
Entrez Gene ID
3236
Gene Name
homeobox D10
Gene Aliases
HOX4, HOX4D, HOX4E, Hox-4.4
Chromosome Location
Chr. 2: 176116764 - 176119942 on Build GRCh38
UniGene ID
Hs.123070
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002148.3 NP_002139.2 1, 2 825
GenBank mRNA BC069619.1 - 1, 2 805
BC074760.2 - 1, 2 804 30915219
BX114645.1 - 1, 2 193
X59373.1 - 1, 2 788

View Full Product Details