Gene Transcripts

Gene Symbol
HSPA1A
Entrez Gene ID
3303
Gene Name
heat shock protein family A (Hsp70) member 1A
Gene Aliases
HEL-S-103, HSP70-1, HSP70-1A, HSP70.1, HSP70I, HSP72, HSPA1
Chromosome Location
Chr. 6: 31815514 - 31817942 on Build GRCh38
UniGene ID
Hs.274402
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005345.5 NP_005336.3 1 2218
GenBank mRNA AB209818.1 - 1 2179
AK096017.1 - 2 1694
AK225692.1 - 1 2189
AK225699.1 - 1 2189
AK291295.1 - 1 2189
AK294182.1 - 2 1982
AK298056.1 - 2 2117
AK298098.1 - 2 1517
AK301243.1 - 2 1898
AK301574.1 - 2 2135
AK303980.1 - 2 1460
AK304618.1 - 2 1955
AK304652.1 - 2 2024
BC002453.2 - 1 2152 3345864
FJ224292.1 - 1 2464

View Full Product Details