Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002542.5 | NP_002533.1 | 2 | 518 | |
NM_016819.3 | NP_058212.1 | 2 | 518 | ||
NM_016820.3 | NP_058213.1 | 2 | 518 | ||
NM_016821.2 | NP_058214.1 | 2 | 518 | ||
NM_016826.2 | NP_058434.1 | 2 | 518 | ||
NM_016827.2 | NP_058436.1 | 2 | 518 | ||
NM_016828.2 | NP_058437.1 | 2 | 518 | ||
NM_016829.2 | NP_058438.1 | 2 | 518 | ||
XM_011533760.1 | XP_011532062.1 | 2 | 513 | ||
XM_017006493.1 | XP_016861982.1 | 2 | 513 | ||
XM_017006494.1 | XP_016861983.1 | 2 | 513 | ||
XM_017006495.1 | XP_016861984.1 | 2 | 513 | ||
XM_017006496.1 | XP_016861985.1 | 2 | 513 | ||
XM_017006497.1 | XP_016861986.1 | 2 | 513 | ||
XM_017006498.1 | XP_016861987.1 | 2 | 513 | ||
XM_017006499.1 | XP_016861988.1 | 2 | 513 | ||
GenBank mRNA | AB000410.1 | - | 2 | 443 | |
AF003595.1 | - | 2 | 231 | ||
AF026691.1 | - | 2 | 382 | ||
AK289858.1 | - | 2 | 471 | ||
BC000657.2 | - | 2 | 472 | 3350168 | |
BM847736.1 | - | 2 | 519 | ||
U88527.1 | - | 2 | 485 | ||
U88620.1 | - | 2 | 485 | ||
U96710.1 | - | 2 | 232 | ||
Y11731.2 | - | 2 | 483 | ||
Y11838.1 | - | 2 | 482 | ||
Y13277.1 | - | 2 | 479 |