Gene Transcripts

Gene Symbol
KRTAP17-1
Entrez Gene ID
83902
Gene Name
keratin associated protein 17-1
Gene Aliases
KAP17.1, KRTAP16.1, KRTAP17.1
Chromosome Location
Chr. 17: 41314917 - 41315695 on Build GRCh38
UniGene ID
Hs.307009
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031964.1 NP_114170.1 1 373
GenBank mRNA AJ406952.1 - 1 373
BC069078.1 - 1 371
BC069297.1 - 1 373

View Full Product Details