Gene Transcripts

Gene Symbol
Rps15
Entrez Gene ID
29285
Gene Name
ribosomal protein S15
Gene Aliases
Chromosome Location
Chr. 7: 12282134 - 12283573 on Build Rnor_6.0
UniGene ID
Rn.3391
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017151.2 NP_058847.1 3 185
GenBank mRNA AY390387.1 - 2 113
FQ211784.1 - 3 185
FQ217117.1 - 3 186
FQ217388.1 - 3 185
FQ217914.1 - 3 185
FQ220999.1 - 3 185
FQ221086.1 - 3 186
FQ221189.1 - 3 185
FQ221217.1 - 3 186
FQ221800.1 - 3 185
FQ222134.1 - 3 185
FQ222137.1 - 3 186
FQ222200.1 - 3 185
FQ222233.1 - 3 184
FQ222539.1 - 3 185
FQ222610.1 - 3 185
FQ222635.1 - 3 188
FQ222928.1 - 3 185
FQ223014.1 - 2 187
FQ223311.1 - 3 186
FQ223478.1 - 3 188
FQ223811.1 - 3 185
FQ223860.1 - 3 181
FQ224530.1 - 3 185
FQ228367.1 - 3 193
FQ228405.1 - 3 188
M19393.1 - 3 185

View Full Product Details