Gene Transcripts
- Gene Symbol
- Myo1e
- Entrez Gene ID
- 25484
- Gene Name
- myosin IE
- Gene Aliases
- MYR5, Myr3
- Chromosome Location
- Chr. 8: 76644715 - 76840240 on Build Rnor_6.0
- UniGene ID
- Rn.8737
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_173101.1 |
NP_775124.1
|
3 |
483 |
|
GenBank mRNA |
X74815.1 |
- |
3 |
483 |
|
View Full Product Details