Gene Transcripts
- Gene Symbol
- Ncstn
- Entrez Gene ID
- 289231
- Gene Name
- nicastrin
- Gene Aliases
- Chromosome Location
- Chr. 13: 90451046 - 90467256 on Build Rnor_6.0
- UniGene ID
- Rn.51975
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_174864.3 |
NP_777353.1
|
15, 16 |
1888 |
|
XM_006250283.1 |
XP_006250345.1
|
14, 15 |
1743 |
|
XM_008769733.1 |
XP_008767955.1
|
15, 16 |
1873 |
|
XM_008769734.1 |
XP_008767956.1
|
15, 16 |
1830 |
|
XM_008769735.1 |
XP_008767957.1
|
13, 14 |
1591 |
|
GenBank mRNA |
AF510722.1 |
- |
14, 15 |
1672 |
|
AY101378.1 |
- |
15, 16 |
1796 |
|
BC074006.1 |
- |
16, 17 |
2003 |
|
BC167747.1 |
- |
14, 15 |
1743 |
8365089 |
FQ212793.1 |
- |
3, 4 |
291 |
|
View Full Product Details