Gene Transcripts

Gene Symbol
Atp4b
Entrez Gene ID
24217
Gene Name
ATPase, H+/K+ exchanging, beta polypeptide
Gene Aliases
S76401S1
Chromosome Location
Chr. 16: 81136218 - 81145131 on Build Rnor_6.0
UniGene ID
Rn.10033
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012510.2 NP_036642.2 4 561
XM_008771385.1 XP_008769607.1 4 386
GenBank mRNA M35535.1 - 4 561
M55655.1 - 4 397

View Full Product Details