Gene Transcripts

Gene Symbol
Me1
Entrez Gene ID
24552
Gene Name
malic enzyme 1, NADP(+)-dependent, cytosolic
Gene Aliases
MOD1
Chromosome Location
Chr. 8: 94256830 - 94368834 on Build Rnor_6.0
UniGene ID
Rn.3519
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012600.2 NP_036732.2 2, 3 220
XM_008766439.1 XP_008764661.1 2, 3 517
GenBank mRNA FQ228526.1 - 2, 3 251

View Full Product Details