Gene Transcripts

Gene Symbol
Casp1
Entrez Gene ID
25166
Gene Name
caspase 1
Gene Aliases
Ice, Il1bc
Chromosome Location
Chr. 8: 2605743 - 2614637 on Build Rnor_6.0
UniGene ID
Rn.37508
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012762.2 NP_036894.2 1, 2 282
GenBank mRNA D85899.1 - 1, 2 282
U14647.1 - 1, 2 282

View Full Product Details