Gene Transcripts

Gene Symbol
Timp3
Entrez Gene ID
25358
Gene Name
TIMP metallopeptidase inhibitor 3
Gene Aliases
Chromosome Location
Chr. 7: 23543125 - 23594162 on Build Rnor_6.0
UniGene ID
Rn.119634
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012886.2 NP_037018.2 4 627
XM_006241162.2 XP_006241224.1 4 656
GenBank mRNA BC097335.1 - 4 627
U27201.1 - 4 342

View Full Product Details