Gene Transcripts

Gene Symbol
Pfkl
Entrez Gene ID
25741
Gene Name
phosphofructokinase, liver
Gene Aliases
ATP-PFK, PFK-L
Chromosome Location
Chr. 20: 11393853 - 11415889 on Build Rnor_6.0
UniGene ID
Rn.4212
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013190.4 NP_037322.1 4 344
XM_008772798.1 XP_008771020.1 5 447
GenBank mRNA BC061791.1 - 4 354
X58865.1 - 4 344

View Full Product Details