Gene Transcripts

Gene Symbol
Serpinh1
Entrez Gene ID
29345
Gene Name
serpin peptidase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1)
Gene Aliases
Cbp2, Serpinh2
Chromosome Location
Chr. 1: 164301010 - 164308306 on Build Rnor_6.0
UniGene ID
Rn.98199
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017173.1 NP_058869.1 2 294
GenBank mRNA BC086529.1 - 1 185
M69246.1 - 2 294

View Full Product Details